Sodium chloride 99-100.5%, granular, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

7544-06
MK754406CS 1217.78 USD
MK754406
Sodium chloride 99-100.5%, granular, AR®, Macron Fine Chemicals™
Sodium chloride

Granular.

Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (NaCl) (dried basis) 99.5 - 100.5 %
Appearance (white, crystalline granules) Passes Test
Solubility (10 in 50) Passes Test
Identification A Passes Test
Identification B Passes Test
Loss on Drying at 105°C ≤0.5 %
Acidity or Alkalinity Passes Test
Limit of Bromide ≤0.010 %
Calcium and Magnesium (as Ca) ≤0.005 %
Magnesium and Alkaline Earth Metals (as Ca) ≤0.01 %
Phosphate (PO₄) ≤0.0025 %
Barium (Ba) Passes Test
Arsenic (As) ≤0.0001 %
Heavy Metals (as Pb) ≤0.0005 %
Iron (Fe) ≤0.0002 %
Sulfate (SO₄) ≤200 ppm
Iodide (I) Passes Test
Copper (Cu) ≤0.0002 %
Sodium Ferrocyanide Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR