Sodium chloride 99.0-100.5% (dried basis), granular USP, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = R

7532-06 7532-19 7532-20 7532-25 7532-04
MK753228EA 1750.19 USD
MK753228 MK753206 MK753219 MK753220 MK753225 MK753204 JT7532-QM
Sodium chloride 99.0-100.5% (dried basis), granular USP, FCC, Macron Fine Chemicals™
Sodium chloride

Optimize biopharmaceutical fermentation and cell culture productivity and improve the efficiency of your pharmaceutical production and biopharmaceutical processes with our proven-effective vitamins and pharmaceutical process minerals. Both the J.T.Baker® and Macron Fine Chemicals™ brands of vitamins and minerals are available as USP, FCC, NF or ACS certified products.

Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Meets F.C.C. Requirements
Food GMP Manufactured Product
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (NaCl)(USP) (dried basis) 99.0 - 100.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Loss on Drying at 105°C ≤0.5 %
USP - Acidity or Alkalinity Passes Test
USP - Iodide (I) Passes Test
USP - Barium (Ba) Passes Test
USP - Magnesium and Alkaline Earth Metals (as Ca) ≤100 ppm
USP - Arsenic (As) ≤1 ppm
USP - Nitrite (NO₂) (Absorbance Value) ≤0.01
USP - Sulfate (SO₄) ≤200 ppm
USP - Phosphate (PO₄) ≤25 ppm
USP - Iron (Fe) ≤2 ppm
USP - Bromide (Br) ≤100 ppm
USP - Appearance of Solution Passes Test
USP - Ferrocyanide Passes Test
USP - Aluminum (Al) ≤0.2 ppm
USP - Potassium (K) ≤500 ppm
FCC - Assay (as NaCl) (FCC) (ignited basis) 99.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Arsenic (As) ≤1 ppm
FCC - Calcium and Magnesium ≤0.35 %
FCC - Heavy Metals (as Pb) ≤2 ppm
FCC - Loss on Drying at 105°C ≤0.5 %
Solubility (10 in 50) Passes Test
Appearance (white, crystalline granules, slightly hygroscopic)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Must be subjected to further processing during the preparation of
injectable dosage forms.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR