di-Sodium tetraborate decahydrate ≥99.5%, granules, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate

Management of Change (MOC) category = N

7457-06 7457-20
MK745706CS 1764.58 USD
MK745706 MK745720
di-Sodium tetraborate decahydrate ≥99.5%, granules, AR® ACS, Macron Fine Chemicals™
di-Sodium tetraborate decahydrate

Granular.

Formula: Na₂B₄O₇·10H₂O
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 75 °C
Density: 1.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149193
CAS Number: 1303-96-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 13,08662

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Na₂B₄O₇·10H₂O) 99.5 - 105.0 %
ACS - pH of 0.01M Solution at 25°C 9.15 - 9.20
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤5 ppm
CHIRALPAK® - Registered trademark of Daicel Chemical Industries Ltd.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR