di-Sodium tetraborate decahydrate ≥99.5%, granules, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate
Management of Change (MOC) category = N
7457-06
7457-20
MK745706CS
1764.58
USD
MK745706
MK745720
di-Sodium tetraborate decahydrate ≥99.5%, granules, AR® ACS, Macron Fine Chemicals™
di-Sodium tetraborate decahydrate
Granular.
Formula:
Na₂B₄O₇·10H₂O Boiling Pt: 320 °C (1013 hPa) Melting Pt: 75 °C Density: 1.73 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149193 CAS Number: 1303-96-4 EINECS: 215-540-4 REACH: 01-2119490790-32 Merck Index: 13,08662 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (Na₂B₄O₇·10H₂O) | 99.5 - 105.0 % |
ACS - pH of 0.01M Solution at 25°C | 9.15 - 9.20 |
ACS - Insoluble Matter | ≤0.005 % |
ACS - Chloride (Cl) | ≤0.001 % |
ACS - Phosphate (PO₄) | ≤0.001 % |
ACS - Sulfate (SO₄) | ≤0.005 % |
ACS - Calcium (Ca) | ≤0.005 % |
ACS - Heavy Metals (as Pb) | ≤0.001 % |
ACS - Iron (Fe) | ≤5 ppm |
CHIRALPAK® - Registered trademark of Daicel Chemical Industries Ltd. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...