di-Sodium tetraborate decahydrate ≥99% NF, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate
Management of Change (MOC) category = R
7418-26
MK741826EA
2356.89
USD
MK741826
di-Sodium tetraborate decahydrate ≥99% NF, Macron Fine Chemicals™
di-Sodium tetraborate decahydrate
Formula:
Na₂B₄O₇·10H₂O Boiling Pt: 320 °C (1013 hPa) Melting Pt: 75 °C Density: 1.73 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149193 CAS Number: 1303-96-4 EINECS: 215-540-4 REACH: 01-2119490790-32 Merck Index: 13,08662 |
Specification Test Results
Meets N.F. Requirements | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Assay (Na₂B₄O₇·10H₂O) | 99.0 - 105.0 % |
NF - Carbonate and Bicarbonate | Passes Test |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
Appearance (colorless crystals or white crystalline powder) | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...