Sand, (dried basis), washed and dried, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Silicium dioxide, Silicium (IV) oxide, Silicon dioxide

Management of Change (MOC) category = N

7062-06
MK706206CS 1094.51 USD
MK706206
Sand, (dried basis), washed and dried, Macron Fine Chemicals™
Sand

Sand washed and dried.


Caution: For laboratory, research, or manufacturing use.

Formula: SiO₂
MW: 60.08
Melting Pt: 1700
Density: 2.6
Storage Temperature: Ambient
MDL Number: MFCD02100519
CAS Number: 7631-86-9
EINECS: 238-878-4

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Substances Soluble in HCl ≤0.16 %
Appearance (fine, off-white to gray granules)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR