Ethyl 4-hydroxybenzoate 98.0-102.0%, GenAR® NF, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
Management of Change (MOC) category = R
MACR6882-18
JT6882-18CS
1524.25
USD
JT6882-18
MK6882-QM
MK6882-18
Ethyl 4-hydroxybenzoate 98.0-102.0%, GenAR® NF, Macron Fine Chemicals™
Ethyl 4-hydroxybenzoate
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%.
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 297 |
MDL Number:
MFCD00002353 CAS Number: 120-47-8 Merck Index: 13,03869 |
Specification Test Results
Meets J.P. Chemical Specifications | |
GMP Manufactured Product | |
Meets N.F. Requirements | |
Meets B.P. Chemical Specifications | |
Meets E.P. Chemical Specifications | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Assay (C₉H₁₀O₃) | 98.0 - 102.0 % |
NF - Appearance (white, crystalline powder) | Passes Test |
NF - Color of Solution | Passes Test |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
NF - Acidity | Passes Test |
NF - Related Substances - p-Hydroxybenzoic Acid | ≤0.5 % |
NF - Related Substances - Unspecified Impurities (each) | ≤0.5 % |
NF - Related Substances - Total Impurities | ≤1.0 % |
NF - Residue on Ignition | ≤0.1 % |
EP/BP - Assay (C₉H₁₀O₃) | 98.0 - 102.0 % |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Acidity | Passes Test |
EP/BP - Related Substances - Impurity A | ≤0.5 % |
EP/BP - Related Substances - Unspecified Impurities (each) | ≤0.5 % |
EP/BP - Related Substances - Total Impurities | ≤1.0 % |
EP/BP - Sulfated Ash | ≤0.1 % |
JP - Acidity | Passes Test |
JP - Assay | 98.0 - 102.0 % |
JP - Clarity and Color of Solution | Passes Test |
JP - Identification | Passes Test |
JP - Melting Point | 115 - 118 °C |
JP - Heavy Metals | ≤20 ppm |
JP-RelatedSubstances- p-Hydroxybenzoic Acid | ≤0.5 % |
JP-Related Substances-Unspecified Impurities (each) | ≤0.5 % |
JP-Related Substances-Total Impurities | ≤1.0 % |
JP - Residue on Ignition | ≤0.10 % |
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...