Potassium hydrogen carbonate 99.5-101.5% (dried basis), granular FCC, USP, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Management of Change (MOC) category = R
6732-27
MK673227EA
4028.23
USD
MK673227
Potassium hydrogen carbonate 99.5-101.5% (dried basis), granular FCC, USP, Macron Fine Chemicals™
Potassium hydrogen carbonate
Formula:
KHCO₃ Melting Pt: 292 °C Density: 2.17 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011402 CAS Number: 298-14-6 EINECS: 206-059-0 Merck Index: 13,07691 |
Specification Test Results
Food GMP Manufactured Product | |
Meets F.C.C. Requirements | |
GMP Manufactured Product | |
Meets U.S.P Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
Bulk Pharmaceutical Chemical | |
USP - Assay (KHCO₃) (dried basis) | 99.5 - 101.5 % |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Loss on Drying | ≤0.25 % |
USP - Normal Carbonate | ≤2.5 % |
FCC - Assay (KHCO₃) (dried basis) | 99.5 - 101.5 % |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Carbonate | Passes Test |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Loss on Drying | ≤0.25 % |
Arsenic (As) | ≤3 ppm |
Solubility | Passes Test |
Appearance (white granules) | Passes Test |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...