Potassium nitrate 99.0-100.5%, fine crystals, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
6715-04
6715-06
6715-20
MK671504CS
1388.81
USD
MK671504
MK671506
MK671520
Potassium nitrate 99.0-100.5%, fine crystals, Macron Fine Chemicals™
Potassium nitrate
Fine crystals.
Formula:
KNO₃ Melting Pt: 334 °C Density: 2.109 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011409 CAS Number: 7757-79-1 EINECS: 231-818-8 UN: 1486 ADR: 5.1,III REACH: 01-2119488224-35 Merck Index: 13,07733 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (KNO₃) | 99.0 - 100.5 % |
Solubility | Passes Test |
Loss on Drying at 105°C | ≤1.0 % |
Reaction | Passes Test |
Identification A | Passes Test |
Identification B | Passes Test |
Identification for Sodium | Passes Test |
Sodium (Na), For Information Only | |
Chlorate | Passes Test |
Chloride (Cl) | ≤0.058 % |
Arsenic (As) | ≤0.0003 % |
Heavy Metals (as Pb) | ≤0.0020 % |
Lead (Pb) | ≤0.0010 % |
Appearance (white to off-white crystals) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...