Ammonia solution 28.0 - 30.0% (NH₃ basis), AR Select® ACS for trace metal analysis, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

Management of Change (MOC) category = N

6665-14 6665-46 6665-18
MK666514CS 382.98 USD
MK666514 MK666546 MK666518
Ammonia solution 28.0 - 30.0% (NH₃ basis), AR Select® ACS for trace metal analysis, Macron Fine Chemicals™
Ammonia solution

AR Select®—A line of acids specifically developed for trace metal analysis; analyzed for up to 32 metals in the low ppb range. 

Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Density: 0.88…0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (as NH₃) 28.0 - 30.0 %
Appearance Passes Test
ACS - Carbon Dioxide (CO₂) ≤0.002 %
ACS - Pyridine Passes Test
ACS - Residue after Ignition ≤0.002 %
ACS - Substances Reducing Permanganate Passes Test
ACS - Specific Gravity at 60°/60°F 0.896 - 0.902
Trace Impurities - Arsenic (As) ≤3.000 ppm
Chloride (Cl) ≤0.5 ppm
Trace Impurities - Heavy Metals ≤0.5 ppm
Nitrate (NO₃) ≤2 ppm
Phosphate (PO₄) ≤2 ppm
Total Sulfur (as SO₄) ≤2 ppm
Determine by Flame Photometry & ICP - Aluminum (Al) ≤0.100 ppm
Determine by Flame Photometry & ICP - Barium (Ba) ≤0.100 ppm
Determine by Flame Photometry & ICP - Beryllium (Be) ≤0.010 ppm
Determine by Flame Photometry & ICP - Bismuth (Bi) ≤0.010 ppm
Determine by Flame Photometry & ICP - Boron (B) ≤0.050 ppm
Determine by Flame Photometry & ICP - Cadmium (Cd) ≤0.100 ppm
Determine by Flame Photometry & ICP - Calcium (Ca) ≤0.100 ppm
Determine by Flame Photometry & ICP - Chromium (Cr) ≤0.050 ppm
Determine by Flame Photometry & ICP - Cobalt (Co) ≤0.050 ppm
Determine by Flame Photometry & ICP - Copper (Cu) ≤0.050 ppm
Determine by Flame Photometry & ICP - Gallium (Ga) ≤0.500 ppm
Determine by Flame Photometry & ICP - Germanium (Ge) ≤0.500 ppm
Determine by Flame Photometry & ICP - Gold (Au) ≤0.500 ppm
Determine by Flame Photometry & ICP - Iron (Fe) ≤0.100 ppm
Determine by Flame Photometry & ICP - Lead (Pb) ≤0.2 ppm
Determine by Flame Photometry & ICP - Lithium (Li) ≤0.050 ppm
Determine by Flame Photometry & ICP - Magnesium (Mg) ≤0.100 ppm
Determine by Flame Photometry & ICP - Manganese (Mn) ≤0.050 ppm
Determine by Flame Photometry & ICP - Mercury (Hg) ≤0.500 ppm
Determine by Flame Photometry & ICP - Molybdenum (Mo) ≤0.010 ppm
Determine by Flame Photometry & ICP - Nickel (Ni) ≤0.500 ppm
Determine by Flame Photometry & ICP - Potassium (K) ≤0.100 ppm
Determine by Flame Photometry & ICP - Silicon (Si) ≤0.100 ppm
Determine by Flame Photometry & ICP - Silver (Ag) ≤0.050 ppm
Determine by Flame Photometry & ICP - Sodium (Na) ≤0.500 ppm
Determine by Flame Photometry & ICP - Strontium (Sr) ≤0.100 ppm
Determine by Flame Photometry & ICP - Thallium (Tl) ≤0.050 ppm
Determine by Flame Photometry & ICP - Tin (Sn) ≤0.100 ppm
Determine by Flame Photometry & ICP - Vanadium (V) ≤0.010 ppm
Determine by Flame Photometry & ICP - Zinc (Zn) ≤0.100 ppm
Determine by Flame Photometry & ICP - Zirconium (Zr) ≤0.010 ppm
Determine by Flame Photometry & ICP

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR