Nitric acid 68.0 - 70.0%, AR Select® ACS for trace metal analysis, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

6623-14 6623-46
MK662314CS 974.55 USD
MK662314 MK662346
Nitric acid 68.0 - 70.0%, AR Select® ACS for trace metal analysis, Macron Fine Chemicals™
Nitric acid

AR Select—A line of acids specifically developed for trace metal analysis; analyzed for up to 32 metals in the low ppb range. 

Formula: HNO₃
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HNO₃) 68.0 - 70.0 %
Appearance Passes Test
ACS - Color (APHA) ≤ 10
ACS - Arsenic (As) ≤ 0.010 ppm
ACS - Arsenic and Antimony (as As) ≤ 0.005 ppm
ACS - Chloride (Cl) ≤ 0.04 ppm
ACS - Heavy Metals (as Pb) ≤ 0.05 ppm
ACS - Residue after Ignition ≤ 5 ppm
ACS - Sulfate (SO₄) ≤ 0.4 ppm
Determine by Flame Photometry & ICP - Aluminum (Al) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Barium (Ba) ≤ 1.0 ppb
Determine by Flame Photometry & ICP - Beryllium (Be) ≤ 1.0 ppb
Determine by Flame Photometry & ICP - Bismuth (Bi) ≤ 10 ppb
Determine by Flame Photometry & ICP - Boron (B) ≤ 10.0 ppb
Determine by Flame Photometry & ICP - Cadmium (Cd) ≤ 1.0 ppb
Determine by Flame Photometry & ICP - * Calcium (Ca) ≤ 200.0 ppb
Determine by Flame Photometry & ICP - Chromium (Cr) ≤ 10.0 ppb
Determine by Flame Photometry & ICP - Cobalt (Co) ≤ 5.0 ppb
Determine by Flame Photometry & ICP - Copper (Cu) ≤ 1.0 ppb
Determine by Flame Photometry & ICP - Gallium (Ga) ≤ 20.0 ppb
Determine by Flame Photometry & ICP - Germanium (Ge) ≤ 10.0 ppb
Determine by Flame Photometry & ICP - Gold (Au) ≤ 10.0 ppb
Determine by Flame Photometry & ICP - Iron (Fe) ≤ 40.0 ppb
Determine by Flame Photometry & ICP - Lead (Pb) ≤ 1.0 ppb
Determine by Flame Photometry & ICP - Lithium (Li) ≤ 5.0 ppb
Determine by Flame Photometry & ICP - Magnesium (Mg) ≤ 10.0 ppb
Determine by Flame Photometry & ICP - Manganese (Mn) ≤ 1.0 ppb
Determine by Flame Photometry & ICP - Mercury (Hg) ≤ 1.0 ppb
Determine by Flame Photometry & ICP - Molybdenum (Mo) ≤ 5.0 ppb
Determine by Flame Photometry & ICP - Nickel (Ni) ≤ 5.0 ppb
Determine by Flame Photometry & ICP - Potassium (K) ≤ 20.0 ppb
Determine by Flame Photometry & ICP - * Silicon (Si) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Silver (Ag) ≤ 5.0 ppb
Determine by Flame Photometry & ICP - * Sodium (Na) ≤ 500.0 ppb
Determine by Flame Photometry & ICP - Strontium (Sr) ≤ 1.0 ppb
Determine by Flame Photometry & ICP - Thallium (Tl) ≤ 20.0 ppb
Determine by Flame Photometry & ICP - Tin (Sn) ≤ 10.0 ppb
Determine by Flame Photometry & ICP - Vanadium (V) ≤ 1.0 ppb
Determine by Flame Photometry & ICP - Zinc (Zn) ≤ 5.0 ppb
Determine by Flame Photometry & ICP - Zirconium (Zr) ≤ 10 ppb
Determine by Flame Photometry & ICP
* May change over time due to extraction from glass container.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR