D-(-)-Mannitol, powder, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
6209-05
MACR6209-04
MK620905EA
1649.43
USD
MK620905
MK6209-04
D-(-)-Mannitol, powder, AR® ACS, Macron Fine Chemicals™
D(-)-Mannitol
Powder.
Formula:
HOH₂C(CH(OH))₄CH₂OH Boiling Pt: 290…295 °C (4 hPa) Melting Pt: 164…169 °C Density: 1.49 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064287 CAS Number: 69-65-8 EINECS: 200-711-8 Merck Index: 13,05769 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Heavy Metals (as Pb) | ≤5 ppm |
ACS - Insoluble Matter | ≤0.01 % |
ACS - Loss on Drying at 105°C | ≤0.05 % |
ACS - Reducing Sugars | Passes Test |
ACS - Residue after Ignition | ≤0.01 % |
ACS - Titrable Acid (meq/g) | ≤0.0008 |
ACS - Specific Rotation [Å]²⁵^D (+) | 23.3 - 24.3 Degree |
ACS - Melting Point | 166 - 168 °C |
Solubility (2 in 20) | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...