Methyl ethyl ketone ≥99%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: EK 500 Additive, Butan-2-one, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

Management of Change (MOC) category = N

6206-08
MK620608CS 1421.38 USD
MK620608
Methyl ethyl ketone ≥99%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Methyl ethyl ketone

For use as an HPLC and TLC solvent. Filtered through a 0.2µm element.

Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.8054 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃COCH₂CH₃) (by GC) ≥99.0 %
ACS - Color ≤15
ACS - Residue after Evaporation ≤5 ppm
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.08 %
Optical Absorbance (1-cm path vs water) - 329 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 335 nm ≤0.50
Optical Absorbance (1-cm path vs water) - 344 nm ≤0.05
Optical Absorbance (1-cm path vs water) - 350 - 400 nm ≤0.01
Titrable Acid (meq/g) ≤0.0005
Filtered through a 0.2 micron filter.
Appearance (clear, colorless liquid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR