Manganese(II) sulfate monohydrate ≥98%, powder, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

6192-20
MK619220EA 3149.21 USD
MK619220
Manganese(II) sulfate monohydrate ≥98%, powder, AR® ACS, Macron Fine Chemicals™
Manganese (II) sulphate monohydrate

Powder.

Formula: MnSO₄·1H₂O
Melting Pt: 700 °C
Density: 2.95 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
ADR: 9,III
Merck Index: 13,05763

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (MnSO₄·H₂O) 98.0 - 101.0 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Chloride (Cl) ≤0.005 %
ACS - Heavy Metals (as Pb) ≤0.002 %
ACS - Insoluble Matter ≤0.01 %
ACS - Iron (Fe) ≤0.002 %
ACS - Loss on Ignition 10.0 - 12.0 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Nickel (Ni) ≤0.02 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.05 %
Solubility Passes Test
ACS - Substances Reducing KMnO₄ Passes Test
ACS - Zinc (Zn) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR