Petroleum spirit, 35…60 °C, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

Management of Change (MOC) category = N

6128-19
MK612819EA 540.28 USD
MK612819
Petroleum spirit, 35…60 °C, Macron Fine Chemicals™
Petroleum spirit
Boiling Pt: 35…60 °C (1013 hPa)
Density: 0.7
Flash Pt: -10
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Stearin Residue Test (increase in weight in g) ≤0.003
Appearance and Color Passes Test
Odor... Passes Test
Boiling Range(Lower) 35 - 60 °C
Boiling Range(Upper) 35 - 60 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR