Magnesium chloride hexahydrate 98.0-101.0% USP, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = HR

5956-88 5956-06 5956-12 5956-40
MK595688EA 2470.41 USD
MK595688 MK595606 MK595612 MK595640 JT5956-QM
Magnesium chloride hexahydrate 98.0-101.0% USP, Macron Fine Chemicals™
Magnesium chloride hexahydrate

Crystals. GMP manufactured.


Caution: Not intended for use in Hemodialysis.

Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (MgCl₂·6H₂O) 98.0 - 101.0 %
USP - Barium (Ba) Passes Test
USP - Calcium (Ca) ≤0.01 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Matter ≤0.005 %
USP - pH of Aqueous Solution 4.5 - 7.0
USP - Potassium (K) Passes Test
USP - Sulfate (SO₄) ≤0.005 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Not intended for use in Hemodialysis.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR