Magnesium chloride hexahydrate 98.0-101.0% USP, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Management of Change (MOC) category = HR
5956-88
5956-06
5956-12
5956-40
MK595688EA
2470.41
USD
MK595688
MK595606
MK595612
MK595640
JT5956-QM
Magnesium chloride hexahydrate 98.0-101.0% USP, Macron Fine Chemicals™
Magnesium chloride hexahydrate
Crystals. GMP manufactured.
Caution: Not intended for use in Hemodialysis.
Formula:
MgCl₂·6H₂O Boiling Pt: 1412 °C (1013 hPa) Melting Pt: 117 °C Density: 1.57 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149781 CAS Number: 7791-18-6 EINECS: 232-094-6 Merck Index: 13,05684 |
Specification Test Results
Meets U.S.P Requirements | |
GMP Manufactured Product | |
Preserve in Tight Containers | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Assay (MgCl₂·6H₂O) | 98.0 - 101.0 % |
USP - Barium (Ba) | Passes Test |
USP - Calcium (Ca) | ≤0.01 % |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Insoluble Matter | ≤0.005 % |
USP - pH of Aqueous Solution | 4.5 - 7.0 |
USP - Potassium (K) | Passes Test |
USP - Sulfate (SO₄) | ≤0.005 % |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. | |
Not intended for use in Hemodialysis. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...