4-Methyl-2-pentanone ≥99% (by GC, corrected for water content), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

5923-08
MK592308CS 1939.92 USD
MK592308
4-Methyl-2-pentanone ≥99% (by GC, corrected for water content), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
4-Methyl-2-pentanone

For use as an HPLC and TLC solvent. Filtered through a 0.2µm element.

Formula: (CH₃)₂CHCH₂COCH₃
Boiling Pt: 117.4 °C (1013 hPa)
Melting Pt: –84 °C
Density: 0.801 g/cm³ (20 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Color ≤15
ACS - Assay (CH₃COCH₂CH(CH₃)₂) (by GC, corrected for water) ≥99 %
ACS - Residue after Evaporation ≤5 ppm
ACS - Titrable Acid (meq/g) ≤0.002
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.08 %
Optical Absorbance (1-cm path vs water) - 335 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 340 nm ≤0.50
Optical Absorbance (1-cm path vs water) - 350 nm ≤0.25
Optical Absorbance (1-cm path vs water) - 360 nm ≤0.15
Optical Absorbance (1-cm path vs water) - 400 nm ≤0.01
Filtered through a 0.2 micron filter.
Product Information (not specifications):
Appearance (Clear. colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR