Sodium thiosulfate 0.20 N (N/5), StandARd® volumetric solution, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

5531-60
MK553160EA 67.24 USD
MK553160
Sodium thiosulfate 0.20 N (N/5), StandARd® volumetric solution, Macron Fine Chemicals™
Sodium thiosulphate
Formula: Na₂S₂O₃
Density: 1.06 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.199 - 0.201


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR