Ferric pyrophosphate soluble, pearls, purified, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Iron(III) pyrophosphate, Iron (III) pyrophosphate

Management of Change (MOC) category = R

MACR5528-25
JT5528-25CS 10994.7 USD
JT5528-25 MK5528-25
Ferric pyrophosphate soluble, pearls, purified, Macron Fine Chemicals™
Ferric pyrophosphate
Formula: Fe₄O₂₁P₆
MW: 745.2100 g/mol
MDL Number: MFCD00016091
CAS Number: 10058-44-3

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (Fe) 10.5 - 12.5 %
Solubility Passes Test
Identification A Passes Test
Identification B Passes Test
Ammonium Salts Passes Test
Arsenic (As) ≤0.0008 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR