N,N-Dimethylacetamide ≥99.0% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

Management of Change (MOC) category = N

5407-08
MK540708CS 2308.78 USD
MK540708
N,N-Dimethylacetamide ≥99.0% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
N,N-Dimethylacetamide

For use as an HPLC and TLC Solvent. Filtered through a 2µm element.

Formula: CH₃CON(CH₃)₂
Boiling Pt: 166.1 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0.937 g/cm³ (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 2810
ADR: 6.1,III
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃CON(CH₃)₂) (by GC) ≥99.0 %
ACS - Residue after Evaporation ≤5 ppm
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.03 %
Optical Absorbance (1-cm path vs water) - 270 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 280 nm ≤0.30
Optical Absorbance (1-cm path vs water) - 290 nm ≤0.15
Optical Absorbance (1-cm path vs water) - 310 nm ≤0.05
Optical Absorbance (1-cm path vs water) - 320 nm ≤0.03
Optical Absorbance (1-cm path vs water) - 360 - 400 nm ≤0.01
Filtered through a 0.2 micron filter.
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR