tert-Butyl methyl ether ≥99.0% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

5398-08
MK539808CS 1490.29 USD
MK539808
tert-Butyl methyl ether ≥99.0% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
tert-Butyl methyl ether

For HPLC.

Formula: (CH₃)₃COCH₃
Boiling Pt: 55.2 °C (1013 hPa)
Melting Pt: –108.6 °C
Density: 0.7405 g/cm³ (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (by GC, corrected for water) ≥99.0 %
ACS - Color (APHA) ≤10
ACS - Peroxide (as H₂O₂) ≤1 ppm
ACS - Residue after Evaporation ≤2 ppm
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Optical Absorbance (1-cm path vs water) - 210 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 225 nm ≤0.50
Optical Absorbance (1-cm path vs water) - 250 nm ≤0.10
Optical Absorbance (1-cm path vs water) - 300 nm ≤0.005
Optical Absorbance (1-cm path vs water) - 350 nm ≤0.005
Optical Absorbance (1-cm path vs water) - 400 nm ≤0.005
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR