1-Propanol ≥99.5% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

5351-08
MK535108CS 1418.82 USD
MK535108
1-Propanol ≥99.5% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
1-Propanol
Formula: H₃CCH₂CH₂OH
Boiling Pt: 97 °C (1013 hPa)
Melting Pt: –127 °C
Density: 0.8053 g/cm³ (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,III
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃(CH₂)₂OH) (by GC, corrected for water) ≥99.5 %
Residue (mg/L) ≤3
Water (H₂O)(by Karl Fischer titrn) ≤0.08 %
Optical Absorbance (1-cm path vs water) - 210 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 225 nm ≤0.50
Optical Absorbance (1-cm path vs water) - 250 nm ≤0.05
Optical Absorbance (1-cm path vs water) - 300 nm ≤0.005
Optical Absorbance (1-cm path vs water) - 400 nm ≤0.005
Appearance (clear, colorless liquid)
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR