n-Heptane ≥99% (by GC), AR®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = N
5177-16
5177-22
5177-02
5177-26
MK517716CS
1117.45
USD
MK517716
MK517722
MK517702
MK517726
n-Heptane ≥99% (by GC), AR®, Macron Fine Chemicals™
n-Heptane
Formula:
H₃C(CH₂)₅CH₃ Boiling Pt: 98.4 °C (1013 hPa) Melting Pt: –90.6 °C Density: 0.6838 g/cm³ (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 EINECS: 205-563-8 UN: 1206 ADR: 3,II Merck Index: 13,04679 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (CH₃(CH₂)₅CH₃) (by GC, corrected for water) | ≥99.0 % |
Acidity (as CH₃COOH) | Passes Test |
Residue after Evaporation | ≤0.001 % |
Sulfur Compounds (as S) | ≤0.005 % |
Specific Gravity at 60°/60°F | 0.68 - 0.72 |
Thiophene | Passes Test |
Appearance (clear, colorless, volatile liquid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...