Heptane (mixture of isomers) ≥99% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

5139-06 5139-10
MK513906CS 1214.67 USD
MK513906 MK513910
Heptane (mixture of isomers) ≥99% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Heptane (mixture of isomers)

For HPLC and TLC applications.

Formula: H₃C(CH₂)₅CH₃
Boiling Pt: 98.4 °C (1013 hPa)
Melting Pt: –90.6 °C
Density: 0.68 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dabde8793eb27ef796098b842bf18b9ced8539b8&vid=d30a8083280d1915a43f51d93c49f9b76eabd579&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (sum of n-C₇H₁₆ and other C₇ hydrocarbons)(by GC, corrected for water) ≥99 %
ACS - Assay (CH₃(CH₂)₅CH₃) (by GC, corrected for water) ≥96 %
ACS - Acidity (as CH₃COOH) Passes Test
ACS - Residue after Evaporation ≤0.0003 %
ACS - Water (H₂O) ≤0.02 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 197 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤0.40
Ultraviolet Absorbance (1.00-cm cell vs. water) - 225 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.01
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR