Petroleum spirit, 35…60 °C, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

Management of Change (MOC) category = N

4983-10
MK498310CS 1243.64 USD
MK498310
Petroleum spirit, 35…60 °C, AR® ACS, Macron Fine Chemicals™
Petroleum spirit
Boiling Pt: 35…60 °C (1013 hPa)
Density: 0.7
Flash Pt: -10
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Acidity Passes Test
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.001 %
Appearance and Color Passes Test
ACS - Boiling Range(Lower) 35 - 60 °C
ACS - Boiling Range(Upper) 35 - 60 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR