Petroleum spirit, 30…75 °C, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

Management of Change (MOC) category = N

4976-08
MK497608CS 1171.89 USD
MK497608
Petroleum spirit, 30…75 °C, AR®, Macron Fine Chemicals™
Petroleum spirit
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Acidity Passes Test
Appearance and Color Passes Test
Residue after Evaporation ≤0.0010 %
Sulfur Compounds (doctor test) Passes Test
Boiling Range(Lower) 30.0 - 75.0 °C
Boiling Range(Upper) 30.0 - 75.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR