L(+)-Lysine monohydrochloride 98.5-101.5% (dried basis), GenAR® USP, Multi-Compendial, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl
MACR4933-09
4933-06
MK4933-09EA
14027.23
USD
MK4933-09
MK4933-06
L(+)-Lysine monohydrochloride 98.5-101.5% (dried basis), GenAR® USP, Multi-Compendial, Macron Fine Chemicals™
L(+)-Lysine monohydrochloride
Formula:
H₂N(CH₂)₄CH(NH₂)CO₂H·HCl MW: 182.65 g/mol |
MDL Number:
MFCD00064564 CAS Number: 657-27-2 |
Specification Test Results
Meets J.P. Chemical Specifications | |
Meets E.P. Chemical Specifications | |
Meets U.S.P Requirements | |
GMP Manufactured Product | |
Meets B.P. Chemical Specifications | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Assay (C₆H₁₄N₂O₂·HCl) (dried basis) | 98.5 - 101.5 % |
USP - Identification | Passes Test |
USP - Specific Rotation [Å]²⁵ ^D (+) | 20.4 - 21.4 Degree |
USP - Loss on Drying at 105°C | ≤0.4 % |
USP - Residue on Ignition | ≤0.1 % |
USP - Sulfate (SO₄) | ≤0.03 % |
USP - Iron (Fe) | ≤0.003 % |
USP - Related Compounds - Individual Impurities | ≤0.5 % |
USP - Related Compounds - Total Impurities | ≤2.0 % |
USP - Chloride (Cl) | 19.0 - 19.6 % |
EP/BP - Assay (C₆H₁₄N₂O₂·HCl) (dried basis) | 98.5 - 101.0 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Identification E | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Specific Rotation [Å]²º^D (+) | 21.0 - 22.5 Degree |
EP/BP-Ninhydrin-Positive Substances-Each | ≤0.2 % |
EP/BP-Ninhydrin-Positive Substances-Total Impurities | ≤1.0 % |
EP/BP - Sulfate (SO₄) | ≤300 ppm |
EP/BP - Ammonium (NH₄) | ≤0.02 % |
EP/BP - Iron (Fe) | ≤30 ppm |
EP/BP - Loss on Drying at 105°C | ≤0.5 % |
EP/BP - Ash (sulfated) | ≤0.1 % |
JP - Assay (C₆H₁₄N₂O₂·HCl) (dried basis) | 98.5 - 101.0 % |
JP - Identification A | Passes Test |
JP - Identification B | Passes Test |
JP - Optical Rotation (+) | 19.0 - 21.5 Degree |
JP - pH (1 in 10) | 5.0 - 6.0 |
JP - Clarity and Color of Solution | Passes Test |
JP - Sulfate (SO₄) | ≤0.028 % |
JP - Ammonium (NH₄) | ≤0.02 % |
JP - Heavy Metals (as Pb) | ≤10 ppm |
JP - Arsenic (As) | ≤2 ppm |
JP - Related Substances | Passes Test |
JP - Loss on Drying at 105°C | ≤1.0 % |
JP - Residue on Ignition | ≤0.10 % |
Endotoxin Concentration, IU/mg, For Information Only | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP (232), EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...