EDTA disodium salt dihydrate ≥99%, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

Management of Change (MOC) category = N

4931-02 4931-20 MACR4931-04 4931-24
MK493102CS 1978.47 USD
MK493102 MK493120 MK4931-04 MK4931-24
EDTA disodium salt dihydrate ≥99%, AR® ACS, Macron Fine Chemicals™
EDTA disodium salt dihydrate

(Ethylenedinitrilo)Tetraacetic Acid, Disodium Salt, Dihydrate powder.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₁₀H₁₄N₂Na₂O₈·2H₂O) 99.0 - 101.0 %
Identification A Passes Test
Identification B Passes Test
Identification C Passes Test
ACS - Heavy Metals (as Pb) ≤0.005 %
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤0.01 %
ACS - Nitrilotriacetic Acid (by polarography) ≤0.1 %
ACS - pH of 5% Solution at 25°C 4.0 - 6.0
Certificate Provided Reports Actual Lot Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR