N,N-Dimethylformamide ≥99.8% (by GC, corrected for water content), AR® (ACS) ACS analytical reagent, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF

Management of Change (MOC) category = N

4929-04 4929-19 4929-08 4929-24
MK492906CS 849.6 USD
MK492906 MK492904 MK492919 MK492908 MK492924
N,N-Dimethylformamide ≥99.8% (by GC, corrected for water content), AR® (ACS) ACS analytical reagent, Macron Fine Chemicals™
N,N-Dimethylformamide

ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Formula: HCON(CH₃)₂
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HCON(CH₃)₂) (by GC, corrected for water) ≥99.8 %
ACS - Color (APHA) ≤15
ACS - Residue after Evaporation ≤0.005 %
ACS - Titrable Acid (meq/g) ≤0.0005
ACS - Titrable Base (meq/g) ≤0.003
ACS - Water (H₂O) ≤0.15 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR