Copper(II) bromide 50% in aqueous solution, AR®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Cupric bromide, Cuprous bromide
Management of Change (MOC) category = N
MACR4895-22
MK4895-22EA
1882.48
USD
MK4895-22
Copper(II) bromide 50% in aqueous solution, AR®, Macron Fine Chemicals™
Copper (II) bromide
Formula:
Br₂Cu MW: 223.35 g/mol Boiling Pt: 900 Melting Pt: 498 Density: 1.50 kg (1 L) |
MDL Number:
MFCD00010970 CAS Number: 7789-45-9 UN: 3264 ADR: 8 II Merck Index: 13,02656 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Appearance (dark brown/black solution) | Passes Test |
Assay (CuBr₂) | 49 - 51 % |
Insoluble Matter | ≤0.005 % |
Sulfate (SO₄) | ≤0.005 % |
Chloride (Cl) | ≤0.15 % |
Substances not Precipitated by H₂S (as SO₄) | ≤0.10 % |
Iron (Fe) | ≤0.003 % |
pH of 10% solution | 2.7 - 3.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...