Dichloromethane ≥99.5% (by GC), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methane dichloride, Methylene chloride, Methylene chloride

Management of Change (MOC) category = N

4884-10
MK488410CS 1802.35 USD
MK488410
Dichloromethane ≥99.5% (by GC), AR® ACS, Macron Fine Chemicals™
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.32 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 203-625-9
UN: 1593
ADR: 6.1,III
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₂Cl₂) (by GC, corrected for water) ≥99.5 %
Appearance Passes Test
ACS - Color (APHA) ≤10
ACS - Free Halogens Passes Test
ACS - Residue after Evaporation ≤0.002 %
ACS - Titrable Acid (meq/g) ≤0.0003
ACS - Water (H₂O)(by coulometry) ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR