Dichloromethane ≥99.0%, GenAR® NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

MACR4874-26
JT4874-QMCS 499.81 USD
JT4874-QM MK4874-26
Dichloromethane ≥99.0%, GenAR® NF, Macron Fine Chemicals™
Dichloromethane
Formula: CH₂Cl₂
Density: 1.32 g/cm³
MDL Number: MFCD00000881
CAS Number: 75-09-2
UN: 1593
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets E.P. Chemical Specifications
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay ≥99.0 %
NF - Identification Passes Test
NF - Free Chlorine (Cl₂) Passes Test
NF - Hydrogen Chloride (HCl) ≤0.001 %
NF - Methanol ≤600 ppm
NF - Nonvolatile Residue ≤0.002 %
NF - Specific Gravity at 25°C 1.318 - 1.322
NF - Water ≤0.02 %
1,1-dichloroethene
EP - Appearance Passes Test
EP - Refractive Index 1.423 - 1.425 Degree
EP - Identification B Passes Test
EP - Identification C Passes Test
EP - Free Chlorine Passes Test
EP - Acidity Passes Test
EP - Residue on Evaporation ≤20 ppm
EP - Relative Density @ 20° C 1.320 - 1.332
EP - Water (KF) ≤0.02 %
EP - Ethanol, v/v ≤2.0 %
EP - 2-methylbut-2-ene, v/v ≤300 ppm
EP - Carbon tetrachloride, v/v ≤10 ppm
EP - Chloroform, v/v ≤50 %
EP - Total other impurities v/v ≤0.1 %
Residual Solvents - The Class 1 solvent 1,1-dichloroethene is likely to
be present. The concentration of 1,1-dichloroethene is quantified and
reported for each batch. Methanol and amylene (2-methyl-butylene) are
intentionally added as stabilizers. The concentration of methanol is
reported in the batch results. The concentration of amylene is less
than 40 ppm based on in-process controls. The Class 2 solvents
cyclohexane (less than 20 ppm) and 1,2-dichloroethene (less than 50 ppm) may be present due to their presence in the process raw material. They
are not selectively removed during processing.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR