Cyclohexanone ≥99.8% (by GC), AR® ACS analytical reagent, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Pimelic Ketone

Management of Change (MOC) category = N

4872-04 4872-08
MK487204CS 2777.74 USD
MK487204 MK487208
Cyclohexanone ≥99.8% (by GC), AR® ACS analytical reagent, Macron Fine Chemicals™
Cyclohexanone

ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Formula: C₆H₁₀(=O)
MW: 98.14 g/mol
Boiling Pt: 155.6
Melting Pt: -31
Density: 0.9478 g/cm³ (20 °C)
Flash Pt: 43
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3 III
REACH: 01-2119453616-35
Merck Index: 13,02755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₆H₁₀O) (by GC, corrected for water) ≥99.8 %
Appearance (clear, colorless liquid) Passes Test
ACS - Color (APHA) ≤10
ACS - Acidity (as CH₃COOH) ≤0.010 %
ACS - Residue after Evaporation ≤0.05 %
ACS - Specific Gravity at 25°/25°C 0.940 - 0.950
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR