Zinc stearate 12.5 - 14.0 % RSN, Hyqual®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
MACR4644-32
JT4644-32CS
1291
USD
JT4644-32
MK4644-32
Zinc stearate 12.5 - 14.0 % RSN, Hyqual®, Macron Fine Chemicals™
Zinc stearate
Formula:
C₃₆H₇₀O₄Zn Density: 1.10 g/cm³ Flash Pt: 279.00 °C |
MDL Number:
MFCD00013031 CAS Number: 557-05-1 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (Zn(C₁₈H₃₅O₂)₂) | 12.5 - 14.0 % |
Appearance | Passes Test |
Identification A | Passes Test |
Identification B | ≥54 °C |
Arsenic (As) | ≤1.500 ppm |
Loss on Drying | ≤1.25 % |
Lead (Pb) | ≤0.001 % |
Alkalies and Alkaline Earths (as SO₄) | ≤1.0 % |
Thru U.S. No. 325 Sieve | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...