Molecular sieve 3A (0.3 nm, 3 Å) Grade 564 Mesh 8 - 12, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate

4490-04
MK449004CS 1137.97 USD
MK449004
Molecular sieve 3A (0.3 nm, 3 Å) Grade 564 Mesh 8 - 12, Macron Fine Chemicals™
Molecular sieve 3A (0.3 nm, 3 Å)
Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR