Toluene ≥99.7% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methylbenzene, Methacide, Benzene, methyl, Phenylmethane, Methane, phenyl-, Toluol, FM approval, Methylbenzol

Management of Change (MOC) category = N

4483-10
MK448310CS 965.41 USD
MK448310
Toluene ≥99.7% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Toluene

Especially intended for use as an HPLC and TLC solvent. Filtered through a 0.2µm element.

Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₆H₅CH₃) (by GC) ≥99.7 %
Color (APHA) ≤10
Residue after Evaporation ≤4 ppm
Substances Darkened by H₂SO₄ Passes Test
Sulfur Compounds (as S) ≤0.003 %
Water (H₂O)(by KF, coulometric) ≤0.03 %
Optical Absorbance (1-cm path vs water): Passes Test
Absorbance - 285 nm ≤1.000
Absorbance - 286 nm ≤0.700
Absorbance - 288 nm ≤0.400
Absorbance - 293 nm ≤0.200
Absorbance - 300 nm ≤0.104
Absorbance - 310 nm ≤0.050
Absorbance - 335 nm ≤0.020
Absorbance 350 - 400 nm ≤0.010

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR