Chloroform ≥99.8% (by GC, corrected for water content) stabilised, ChromAR® ACS for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: Stabilised with amylene
Management of Change (MOC) category = N

4443-06 4443-10
MK444306CS 768.06 USD
MK444306 MK444310
Chloroform ≥99.8% (by GC, corrected for water content) stabilised, ChromAR® ACS for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Chloroform

For use as an HPLC and TLC solvent. Stabilized with hydrocarbons. Filtered through a 0.2 µm element.

Formula: CHCl₃
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CHCl₃) (by GC, corrected for water) ≥99.8 %
ACS - Acetone, Aldehydes (as (CH₃)₂CO) Passes Test
ACS - Acid and Chloride Passes Test
ACS - Color (APHA) ≤10
ACS - Free Chlorine Passes Test
ACS - Lead (Pb) ≤0.050 ppm
ACS - Residue after Evaporation ≤0.0003 %
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Suitability for Use in Dithizone Tests Passes Test
ACS - Water (H₂O) ≤0.02 %
ACS - Ultraviolet Absorbance (1.00-cm cell vs. water): Passes Test
ACS UV Absorbance - 245 nm ≤1.00
ACS UV Absorbance - 254 nm ≤0.15
ACS UV Absorbance - 255 nm ≤0.25
ACS UV Absorbance - 260 nm ≤0.15
ACS UV Absorbance - 265 nm ≤0.05
ACS UV Absorbance - 270 nm ≤0.05
ACS Absorbance 280 nm ≤0.01
ACS Absorbance 300 nm ≤0.01
ACS Absorbance - 350 nm ≤0.005
ACS Absorbance - 350 - 400 nm ≤0.01
ACS - Absorbance Passes Test
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR