Chloroform ≥99.8% (by GC), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

4441-10
MK444110CS 1926.37 USD
MK444110
Chloroform ≥99.8% (by GC), AR® ACS, Macron Fine Chemicals™
Chloroform

Clear, colorless liquid.

Formula: CHCl₃
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CHCl₃) (by GC) ≥99.8 %
ACS - Acetone, Aldehydes (as (CH₃)₂CO) Passes Test
ACS - Acid and Chloride Passes Test
ACS - Color (APHA) ≤10
ACS - Free Chlorine Passes Test
ACS - Lead (Pb) ≤0.050 ppm
ACS - Residue after Evaporation ≤0.001 %
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Suitability for Use in Dithizone Tests Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR