Chloroform, purified, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

4432-10
MK443210CS 473.15 USD
MK443210 MK443228
Chloroform, purified, Macron Fine Chemicals™
Chloroform
Formula: CHCl₃
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Acid and Phosgene Passes Test
Aldehydes and Ketones (as HCHO) Passes Test
Chlorinated Decomposition Products and Chloride Passes Test
Free Chlorine Passes Test
Nonvolatile Residue (w/v) ≤0.002 %
Readily Carbonizable Substances Passes Test
Specific Gravity at 25°/25°C 1.476 - 1.480
Water (H₂O) Passes Test
Appearance (clear, colorless liquid)
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR