Chlorobenzene ≥99.5%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

Management of Change (MOC) category = N

4426-08
MK442608CS 2230.62 USD
MK442608
Chlorobenzene ≥99.5%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Chlorobenzene

For use as an HPLC or TLC solvent. Filtered through a 0.2µm element.

Formula: C₆H₅Cl
MW: 112.56 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.11 g/cm³ (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₆H₅Cl) (by GC, corrected for water) ≥99.5 %
ACS - Color (APHA) ≤30
ACS - Residue after Evaporation ≤2 ppm
ACS - Titrable Acid (meq/g) ≤0.004 %
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.03 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 288 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 325 nm ≤0.04
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm ≤0.005
Filtered through a 0.2 micron filter.
Appearance (clear, colorless liquid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR