Imidazole ≥99.0%, OR, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

Management of Change (MOC) category = NI

4337-09
MK433755EA 72.27 USD
MK433755 MK433709
Imidazole ≥99.0%, OR, Macron Fine Chemicals™
Imidazole

Macron Fine Chemicals™ OR: organic reagents that are suitable for research applications.

Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 89…90 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8,III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay ≥99.0 %
Water (H₂O) ≤0.5 %
Appearance (white to off white flakes or crystalline powder)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR