Ammonium sulfide 20.0 - 24.0% in aqueous solution, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

3524-04
MK352404CS 3526.69 USD
MK352404
Ammonium sulfide 20.0 - 24.0% in aqueous solution, AR®, Macron Fine Chemicals™
Ammonium sulphide
Formula: H₈N₂S
MW: 68.14 g/mol
MDL Number: MFCD00010892
CAS Number: 12135-76-1
EINECS: 235-223-4
UN: 2683
ADR: 8 (6.1) (3),II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (as (NH₄)₂S) 20.0 - 24.0 %
Antimony (Sb) ≤0.0005 %
Arsenic (As) ≤0.0001 %
Carbonate (CO₃) ≤0.005 %
Chloride (Cl) ≤0.005 %
Residue after Ignition ≤0.04 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR