Ammonium sulfate 99.0-100.5%, granular FCC, purified, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

Management of Change (MOC) category = RL

3496-28
MK3496-28EA 1834.68 USD
MK3496-28
Ammonium sulfate 99.0-100.5%, granular FCC, purified, Macron Fine Chemicals™
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Density: 1.77 g/cm³
MDL Number: MFCD00003391
CAS Number: 7783-20-2

Order Now

Specification Test Results

Meets F.C.C. Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
FCC - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
FCC - Arsenic (As) ≤3 ppm
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Iron (Fe) ≤0.0010 %
FCC - Lead (Pb) ≤3 mg/kg
FCC - pH of Aqueous Solution 4.5 - 6.0
FCC - Residue after Ignition ≤0.25 %
FCC - Selenium (Se) ≤0.0030 %
Solubility Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR