Ammonium nitrate ≥95%, crystals, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = N
3436-06
3436-20
MK343606CS
1372.33
USD
MK343606
MK343620
JT3436-22
Ammonium nitrate ≥95%, crystals, AR® ACS, Macron Fine Chemicals™
Ammonium nitrate
Crystals.
Formula:
NH₄NO₃ Boiling Pt: 230 °C (1013 hPa) Melting Pt: 169 °C Density: 1.73 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011425 CAS Number: 6484-52-2 EINECS: 229-347-8 UN: 1942 ADR: 5.1,III Merck Index: 13,00538 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (as NH₄NO₃) | ≥95.0 % |
ACS - Chloride (Cl) | ≤5 ppm |
ACS - Heavy Metals (as Pb) | ≤5 ppm |
ACS - Insoluble Matter | ≤0.005 % |
ACS - Iron (Fe) | ≤2 ppm |
ACS - Nitrite (NO₂) | Passes Test |
ACS - pH of 5% Solution at 25°C | 4.5 - 6.0 |
ACS - Phosphate (PO₄) | ≤5 ppm |
ACS - Residue after Ignition | ≤0.01 % |
ACS - Sulfate (SO₄) | ≤0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...