Ammonium heptamolybdate tetrahydrate 81.0-83.0%, crystals, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

3420-04
MK342006CS 7200.69 USD
MK342006 MK342020 MK342004 JT3420-QM MK3420-19
Ammonium heptamolybdate tetrahydrate 81.0-83.0%, crystals, AR® ACS, Macron Fine Chemicals™
Ammonium heptamolybdate tetrahydrate

Crystals.

Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Density: 2.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (as MoO₃) 81.0 - 83.0 %
Appearance Passes Test
ACS - Arsenate, Phosphate and Silicate (as SiO₂) ≤0.001 %
ACS - Chloride (Cl) ≤0.002 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Insoluble Matter ≤0.005 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Nitrate (NO₃) Passes Test
ACS - Potassium (K) ≤0.01 %
ACS - Phosphate (PO₄) ≤0.0005 %
ACS - Sodium (Na) ≤0.01 %
ACS - Sulfate (SO₄) ≤0.02


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR