Ammonia solution 28,0 - 30,0% (NH₃ basis), AR® ACS for trace metal analysis (26° Baume), Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

3256-48 3256-20 3256-45 3256-05 MACR3256-14
MK325648CS 1222.12 USD
MK325648 MK325620 MK325645 MK325605 MK3256-14
Ammonia solution 28,0 - 30,0% (NH₃ basis), AR® ACS for trace metal analysis (26° Baume), Macron Fine Chemicals™
Ammonia solution

ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Formula: NH₃
Boiling Pt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Density: 0.90 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (as NH₃) 28.0 - 30.0 %
Appearance Passes Test
ACS - Carbon Dioxide (CO₂) ≤0.002 %
ACS - Residue after Ignition ≤0.002 %
ACS - Substances Reducing Permanganate Passes Test
Arsenic and Antimony (as As) ≤3 ppm
Chloride (Cl) ≤0.5 ppm
Trace Impurities - Heavy Metals ≤0.5 ppm
Nitrate (NO₃) ≤2 ppm
Phosphate (PO₄) ≤2 ppm
Sulfate (SO₄) ≤2 ppm
Trace Impurities - Aluminum (Al) ≤0.300 ppm
Trace Impurities - Boron (B) ≤0.100 ppm
Trace Impurities - Calcium (Ca) ≤0.300 ppm
Trace Impurities - Chromium (Cr) ≤0.200 ppm
Trace Impurities - Copper (Cu) ≤0.10 ppm
Trace Impurities - Gold (Au) ≤0.300 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm
Trace Impurities - Lead (Pb) ≤0.200 ppm
Trace Impurities - Magnesium (Mg) ≤0.300 ppm
Trace Impurities - Manganese (Mn) ≤0.200 ppm
Trace Impurities - Nickel (Ni) ≤0.1 ppm
Trace Impurities - Potassium (K) ≤0.300 ppm
Trace Impurities - Sodium (Na) ≤2.000 ppm
Trace Impurities - Tin (Sn) ≤0.300 ppm
Trace Impurities - Titanium (Ti) ≤0.300 ppm
Trace Impurities - Zinc (Zn) ≤0.300 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR