Methanol, anhydrous ≥99.9%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol

Management of Change (MOC) category = N

3041-10 3041-06
MK304110CS 877.86 USD
MK304110 MK304106 JT3041-68
Methanol, anhydrous ≥99.9%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Methanol

For use as an HPLC and TLC Solvent. Filtered through a 0.2µm element. Soluble in water.

Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃OH) (by GC) ≥99.9 %
Carbonyl Compounds - Acetaldehyde ≤0.001 %
Carbonyl Compounds - Acetone ≤0.001 %
Carbonyl Compounds - Formaldehyde ≤0.001 %
Color (APHA) ≤10
Residue after Evaporation ≤0.0002 %
Solubility in H₂O Passes Test
Substances Darkened by H₂SO₄ Passes Test
Substances Reducing Permanganate Passes Test
Titrable Acid (meq/g) ≤0.0003
Titrable Base (meq/g) ≤0.0002
Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 205 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤0.80
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.25
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.20
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm ≤0.04
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 - 400 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water): Passes Test
HPLC Gradient Elution (No Peak Greater than 25% of Full Scale)(0.005 au) Passes Test
Appearance (clear, colorless liquid)
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR