2-Propanol ≥99.5%, AR® ACS for histology, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

Management of Change (MOC) category = N

3032-22 3032-02 3032-16 3032-21 3032-06
MK303222CS 228.63 USD
MK303222 MK303202 MK303216 MK303221 MK303206
2-Propanol ≥99.5%, AR® ACS for histology, Macron Fine Chemicals™
2-Propanol

Clear, colorless liquid. Suitable for histological use. Soluble in water.

Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance (clear, colorless liquid) Passes Test
Infrared Spectrum (Conforms to Standard) Passes Test
ACS - Assay (CH₃CHOHCH₃) ≥99.5 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.001 %
ACS - Solubility in H₂O Passes Test
ACS - Titrable Acid or Base (meq/g) ≤0.0001
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.2 %
Carbonyl Compounds - Propionaldehyde ≤0.002 %
Carbonyl Compounds - Acetone ≤0.002 %
Date Received: / /
Date Opened: / /


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR