Methanol absolute, anhydrous ≥99.8% (by GC), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol

Management of Change (MOC) category = N

3016-02 3016-22 3016-25 3016-06 3016-16 3016-R
MK301602CS 894.82 USD
MK301602 MK301622 MK301625 MK301606 MK301616 JT3016-R JT3016-30
Methanol absolute, anhydrous ≥99.8% (by GC), AR® ACS, Macron Fine Chemicals™
Methanol

Low acetone. Soluble in water.

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: -98
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃OH) (by GC) ≥99.8 %
Appearance (clear, colorless liquid) Passes Test
Carbonyl Compounds - Acetaldehyde ≤0.001 %
Carbonyl Compounds - Acetone ≤0.001 %
Carbonyl Compounds - Formaldehyde ≤0.001 %
Color (APHA) ≤10
Heavy Metals (as Pb) ≤0.0001 %
Identification Passes Test
Residue after Evaporation ≤0.001 %
Solubility in H₂O Passes Test
Substances Darkened by H₂SO₄ Passes Test
Substances Reducing Permanganate Passes Test
Titrable Acid (meq/g) ≤0.0003
Titrable Base (meq/g) ≤0.0002
Water (H₂O)(by Karl Fischer titrn) ≤0.1 %
Date Received: / /
Date Opened: / /


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR